missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-iodopyridine 97.0+%, TCI America™

Catalog No. B28775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28775G 5 g
B287725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28775G Supplier TCI America Supplier No. B28775G

Chemical Identifiers

CAS 223463-13-6
Molecular Formula C5H3BrIN
Molecular Weight (g/mol) 283.894
MDL Number MFCD00691506
InChI Key HSNBRDZXJMPDGH-UHFFFAOYSA-N
Synonym 2-iodo-5-bromopyridine, 5-bromo-2-iodo-pyridine, 2-iodo-5-bromo-pyridine, 2-iodo-5-bromo pyridine, pyridine, 5-bromo-2-iodo, 5-bromo-2-iodo pyridine, pubchem2409, 5-cromo-2-iodopyridine, 5-bromo-2-iodopyridine, acmc-209fv4
PubChem CID 593690
IUPAC Name 5-bromo-2-iodopyridine
SMILES C1=CC(=NC=C1Br)I

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 5 g
Formula Weight 283.89
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-iodopyridine