missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-iodopyridine, 98%

CAS: 223463-13-6 | C5H3BrIN | 283.89 g/mol

$110.12 - $110.12

Chemical Identifiers

CAS 223463-13-6
Molecular Formula C5H3BrIN
Molecular Weight (g/mol) 283.89
MDL Number MFCD00691506
InChI Key HSNBRDZXJMPDGH-UHFFFAOYSA-N
Synonym 2-iodo-5-bromopyridine, 5-bromo-2-iodo-pyridine, 2-iodo-5-bromo-pyridine, 2-iodo-5-bromo pyridine, pyridine, 5-bromo-2-iodo, 5-bromo-2-iodo pyridine, pubchem2409, 5-cromo-2-iodopyridine, 5-bromo-2-iodopyridine, acmc-209fv4
PubChem CID 593690
IUPAC Name 5-bromo-2-iodopyridine
SMILES C1=CC(=NC=C1Br)I
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC365940050
SDS View Documents
Thermo Scientific Chemicals
365940050
5 g Glass bottle
Each for $110.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

223463-13-6
283.89
HSNBRDZXJMPDGH-UHFFFAOYSA-N
593690
C1=CC(=NC=C1Br)I
C5H3BrIN
MFCD00691506
2-iodo-5-bromopyridine, 5-bromo-2-iodo-pyridine, 2-iodo-5-bromo-pyridine, 2-iodo-5-bromo pyridine, pyridine, 5-bromo-2-iodo, 5-bromo-2-iodo pyridine, pubchem2409, 5-cromo-2-iodopyridine, 5-bromo-2-iodopyridine, acmc-209fv4
5-bromo-2-iodopyridine

Specifications

223463-13-6
100.0
White to Beige
97.5% min. (GC)
C5H3BrIN
5 g
2-iodo-5-bromopyridine, 5-bromo-2-iodo-pyridine, 2-iodo-5-bromo-pyridine, 2-iodo-5-bromo pyridine, pyridine, 5-bromo-2-iodo, 5-bromo-2-iodo pyridine, pubchem2409, 5-cromo-2-iodopyridine, 5-bromo-2-iodopyridine, acmc-209fv4
C1=CC(=NC=C1Br)I
283.89
283.89
Crystalline Powder
97.5
112°C to 113°C
Authentic
Glass bottle
MFCD00691506
20, II, 154
HSNBRDZXJMPDGH-UHFFFAOYSA-N
5-bromo-2-iodopyridine
593690
98%
5-Bromo-2-iodopyridine, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only