missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-iodopyridine, 98%

Catalog No. AAL1961006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1961006 5 g
AAL1961014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1961006 Supplier Thermo Scientific Chemicals Supplier No. L1961006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 223463-13-6 | C5H3BrIN | 283.894 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 223463-13-6
Molecular Formula C5H3BrIN
Molecular Weight (g/mol) 283.894
MDL Number MFCD00691506
InChI Key HSNBRDZXJMPDGH-UHFFFAOYSA-N
Synonym 2-iodo-5-bromopyridine, 5-bromo-2-iodo-pyridine, 2-iodo-5-bromo-pyridine, 2-iodo-5-bromo pyridine, pyridine, 5-bromo-2-iodo, 5-bromo-2-iodo pyridine, pubchem2409, 5-cromo-2-iodopyridine, 5-bromo-2-iodopyridine, acmc-209fv4
PubChem CID 593690
IUPAC Name 5-bromo-2-iodopyridine
SMILES C1=CC(=NC=C1Br)I

Specifications

Melting Point 113°C to 116°C
Quantity 5 g
Beilstein 2119
Sensitivity Light sensitive
Formula Weight 283.89
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-iodopyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only