missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxypyridine, 97%

Catalog No. p-7028127
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2526706 5 g
AAB2526714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2526706 Supplier Thermo Scientific Chemicals Supplier No. B2526706
Only null left

CAS: 13466-38-1 | C5H4BrNO | 173.997 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13466-38-1
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 173.997
MDL Number MFCD00234042
InChI Key NDMZZQRNZFWMEZ-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxypyridine, 5-bromopyridin-2-ol, 2-hydroxy-5-bromopyridine, 5-bromopyridin-2 1h-one, 5-bromo-2-pyridinol, 5-bromo-2 1h-pyridinone, 5-bromo-2 1h-pyridone, 2 1h-pyridinone, 5-bromo, 3-bromo-6-hydroxypyridine, 5-bromo-1,2-dihydropyridin-2-one
PubChem CID 599528
IUPAC Name 5-bromo-1H-pyridin-2-one
SMILES C1=CC(=O)NC=C1Br

Specifications

Melting Point 178°C to 182°C
Quantity 5 g
Beilstein 108751
Formula Weight 174
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-hydroxypyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only