missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxynicotinic Acid 98.0+%, TCI America™

Catalog No. p-7123127
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B37291G 1 g
B37295G 5 g
2 options

Catalog No. B37291G

Supplier: TCI America B37291G

Only null left

Chemical Identifiers

CAS 104612-36-4
Molecular Formula C6H4BrNO3
Molecular Weight (g/mol) 218.006
MDL Number MFCD07363801
InChI Key GYXOTADLHQJPIP-UHFFFAOYSA-N
Synonym 5-Bromo-2-hydroxy-3-pyridinecarboxylic Acid
PubChem CID 2764024
IUPAC Name 5-bromo-2-oxo-1H-pyridine-3-carboxylic acid
SMILES C1=C(C(=O)NC=C1Br)C(=O)O

Specifications

Melting Point 245°C
Color Yellow
Quantity 1 g
Formula Weight 218.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-hydroxynicotinic Acid