missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxynicotinic Acid 98.0+%, TCI America™

Catalog No. B37295G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37295G 5 g
B37291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37295G Supplier TCI America Supplier No. B37295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 104612-36-4 | C6H4BrNO3 | 218.006 g/mol

Chemical Identifiers

CAS 104612-36-4
Molecular Formula C6H4BrNO3
Molecular Weight (g/mol) 218.006
MDL Number MFCD07363801
InChI Key GYXOTADLHQJPIP-UHFFFAOYSA-N
Synonym 5-Bromo-2-hydroxy-3-pyridinecarboxylic Acid
PubChem CID 2764024
IUPAC Name 5-bromo-2-oxo-1H-pyridine-3-carboxylic acid
SMILES C1=C(C(=O)NC=C1Br)C(=O)O

Specifications

Melting Point 245°C
Color Yellow
Quantity 5 g
Formula Weight 218.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-hydroxynicotinic Acid