missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5'-Bromo-2'-hydroxyacetophenone 97.0+%, TCI America™
$38.51 - $96.26
Chemical Identifiers
| CAS | 1450-75-5 |
|---|---|
| Molecular Formula | C8H7BrO2 |
| Molecular Weight (g/mol) | 215.046 |
| MDL Number | MFCD00191850 |
| InChI Key | HQCCNFFIOWYINW-UHFFFAOYSA-N |
| Synonym | 5'-bromo-2'-hydroxyacetophenone, 1-5-bromo-2-hydroxyphenyl ethanone, 2-acetyl-4-bromophenol, 5-bromo-2-hydroxyacetophenone, 2'-hydroxy-5'-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl ethan-1-one, ethanone, 1-5-bromo-2-hydroxyphenyl, 2-hydroxy-5-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl-ethanon |
| PubChem CID | 95991 |
| IUPAC Name | 1-(5-bromo-2-hydroxyphenyl)ethanone |
| SMILES | CC(=O)C1=C(C=CC(=C1)Br)O |
Chemical Identifiers
| 1450-75-5 | |
| 215.046 | |
| HQCCNFFIOWYINW-UHFFFAOYSA-N | |
| 95991 | |
| CC(=O)C1=C(C=CC(=C1)Br)O |
| C8H7BrO2 | |
| MFCD00191850 | |
| 5'-bromo-2'-hydroxyacetophenone, 1-5-bromo-2-hydroxyphenyl ethanone, 2-acetyl-4-bromophenol, 5-bromo-2-hydroxyacetophenone, 2'-hydroxy-5'-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl ethan-1-one, ethanone, 1-5-bromo-2-hydroxyphenyl, 2-hydroxy-5-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl-ethanon | |
| 1-(5-bromo-2-hydroxyphenyl)ethanone |
Specifications
| 1450-75-5 | |
| White-Yellow | |
| MFCD00191850 | |
| 5'-bromo-2'-hydroxyacetophenone, 1-5-bromo-2-hydroxyphenyl ethanone, 2-acetyl-4-bromophenol, 5-bromo-2-hydroxyacetophenone, 2'-hydroxy-5'-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl ethan-1-one, ethanone, 1-5-bromo-2-hydroxyphenyl, 2-hydroxy-5-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl-ethanon | |
| CC(=O)C1=C(C=CC(=C1)Br)O | |
| 215.046 | |
| 215.05 | |
| Crystalline Powder |
| 58°C | |
| C8H7BrO2 | |
| 5 g | |
| HQCCNFFIOWYINW-UHFFFAOYSA-N | |
| 1-(5-bromo-2-hydroxyphenyl)ethanone | |
| 95991 | |
| ≥97.0% (GC) | |
| 5′-Bromo-2′-hydroxyacetophenone |
Safety and Handling
RTECSNumber : KM5556350
TSCA : No