Learn More
5'-Bromo-2'-hydroxyacetophenone, 98%
CAS: 1450-75-5 | C8H7BrO2 | 215.046 g/mol
$97.26 - $305.40
Chemical Identifiers
| CAS | 1450-75-5 |
|---|---|
| Molecular Formula | C8H7BrO2 |
| Molecular Weight (g/mol) | 215.046 |
| MDL Number | MFCD00191850 |
| InChI Key | HQCCNFFIOWYINW-UHFFFAOYSA-N |
| Synonym | 5'-bromo-2'-hydroxyacetophenone, 1-5-bromo-2-hydroxyphenyl ethanone, 2-acetyl-4-bromophenol, 5-bromo-2-hydroxyacetophenone, 2'-hydroxy-5'-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl ethan-1-one, ethanone, 1-5-bromo-2-hydroxyphenyl, 2-hydroxy-5-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl-ethanon |
| PubChem CID | 95991 |
| IUPAC Name | 1-(5-bromo-2-hydroxyphenyl)ethanone |
| SMILES | CC(=O)C1=C(C=CC(=C1)Br)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6197909
|
Thermo Scientific Chemicals
H6197909 |
10 g |
N/A
|
|
|||||
|
AAH6197918
|
Thermo Scientific Chemicals
H6197918 |
50 g |
N/A
|
|
|||||
Description
5?-Bromo-2?-hydroxyacetophenone (5-Bromo-2-hydroxyacetophenone) may be used in ligand in the preparation of tetrahedral metallocene complexes containing vanadium(IV), synthesis of {2?-[1-(5-bromo-2-oxidophenyl) ethylidene] benzohydrazidato (2-)} tris(pyridine) nickel(II)] pyridine solvate, synthesis of N?-[1-(5-bromo-2-hydroxyphenyl)ethylidene]-3,4,5-trihydroxybenzohydrazide dimethyl sulfoxide solvate trihydrate, preparation of 6-bromochromen-4-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5′-Bromo-2′-hydroxyacetophenone (5-Bromo-2-hydroxyacetophenone) may be used in ligand in the preparation of tetrahedral metallocene complexes containing vanadium(IV), synthesis of {2′-[1-(5-bromo-2-oxidophenyl) ethylidene] benzohydrazidato (2-)} tris(pyridine) nickel(II)] pyridine solvate, synthesis of N′-[1-(5-bromo-2-hydroxyphenyl)ethylidene]-3,4,5-trihydroxybenzohydrazide dimethyl sulfoxide solvate trihydrate, preparation of 6-bromochromen-4-one.
Solubility
Soluble in Chloroform
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 1450-75-5 | |
| 215.046 | |
| HQCCNFFIOWYINW-UHFFFAOYSA-N | |
| 95991 | |
| CC(=O)C1=C(C=CC(=C1)Br)O |
| C8H7BrO2 | |
| MFCD00191850 | |
| 5'-bromo-2'-hydroxyacetophenone, 1-5-bromo-2-hydroxyphenyl ethanone, 2-acetyl-4-bromophenol, 5-bromo-2-hydroxyacetophenone, 2'-hydroxy-5'-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl ethan-1-one, ethanone, 1-5-bromo-2-hydroxyphenyl, 2-hydroxy-5-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl-ethanon | |
| 1-(5-bromo-2-hydroxyphenyl)ethanone |
Specifications
| 1450-75-5 | |
| White to Yellow | |
| MFCD00191850 | |
| 5'-bromo-2'-hydroxyacetophenone, 1-5-bromo-2-hydroxyphenyl ethanone, 2-acetyl-4-bromophenol, 5-bromo-2-hydroxyacetophenone, 2'-hydroxy-5'-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl ethan-1-one, ethanone, 1-5-bromo-2-hydroxyphenyl, 2-hydroxy-5-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl-ethanon | |
| HQCCNFFIOWYINW-UHFFFAOYSA-N | |
| 1-(5-bromo-2-hydroxyphenyl)ethanone | |
| 95991 | |
| 98% | |
| 5'-Bromo-2'-hydroxyacetophenone |
| 56°C to 60°C | |
| C8H7BrO2 | |
| 10 g | |
| Soluble in Chloroform | |
| CC(=O)C1=C(C=CC(=C1)Br)O | |
| 215.046 | |
| 215.04 | |
| Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : KM5556350
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only