missing translation for 'onlineSavingsMsg'
Learn More

5'-Bromo-2'-hydroxyacetophenone, 98%

Catalog No. AAH6197918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6197918 50 g
AAH6197909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6197918 Supplier Thermo Scientific Chemicals Supplier No. H6197918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1450-75-5 | C8H7BrO2 | 215.046 g/mol

5?-Bromo-2?-hydroxyacetophenone (5-Bromo-2-hydroxyacetophenone) may be used in ligand in the preparation of tetrahedral metallocene complexes containing vanadium(IV), synthesis of {2?-[1-(5-bromo-2-oxidophenyl) ethylidene] benzohydrazidato (2-)} tris(pyridine) nickel(II)] pyridine solvate, synthesis of N?-[1-(5-bromo-2-hydroxyphenyl)ethylidene]-3,4,5-trihydroxybenzohydrazide dimethyl sulfoxide solvate trihydrate, preparation of 6-bromochromen-4-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5′-Bromo-2′-hydroxyacetophenone (5-Bromo-2-hydroxyacetophenone) may be used in ligand in the preparation of tetrahedral metallocene complexes containing vanadium(IV), synthesis of {2′-[1-(5-bromo-2-oxidophenyl) ethylidene] benzohydrazidato (2-)} tris(pyridine) nickel(II)] pyridine solvate, synthesis of N′-[1-(5-bromo-2-hydroxyphenyl)ethylidene]-3,4,5-trihydroxybenzohydrazide dimethyl sulfoxide solvate trihydrate, preparation of 6-bromochromen-4-one.

Solubility
Soluble in Chloroform

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1450-75-5
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00191850
InChI Key HQCCNFFIOWYINW-UHFFFAOYSA-N
Synonym 5'-bromo-2'-hydroxyacetophenone, 1-5-bromo-2-hydroxyphenyl ethanone, 2-acetyl-4-bromophenol, 5-bromo-2-hydroxyacetophenone, 2'-hydroxy-5'-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl ethan-1-one, ethanone, 1-5-bromo-2-hydroxyphenyl, 2-hydroxy-5-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl-ethanon
PubChem CID 95991
IUPAC Name 1-(5-bromo-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)Br)O

Specifications

Melting Point 56°C to 60°C
Color White to Yellow
Quantity 50 g
Solubility Information Soluble in Chloroform
Formula Weight 215.04
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5'-Bromo-2'-hydroxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber KM5556350
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only