missing translation for 'onlineSavingsMsg'
Learn More

5'-Bromo-2'-hydroxy-3'-nitroacétophénone 98,0+%, TCI America™

Numéro de catalogue. B45565G
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B45565G 5 g
B455625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B45565G Fournisseur TCI America Code fournisseur B45565G

Identifiants chimiques

CAS 70978-54-0
Molecular Formula C8H6BrNO4
Molecular Weight (g/mol) 260.04
MDL Number MFCD01631134
InChI Key CLNIBJASCGZXHH-UHFFFAOYSA-N
Synonym 1-5-bromo-2-hydroxy-3-nitrophenyl ethanone, 5'-bromo-2'-hydroxy-3'-nitroacetophenone, 3-bromo-6-hydroxy-5-nitroacetophenone, 5'-bromo-3'-nitro-2'-hydroxyacetophenone, 5-bromo-2-hydroxy-3-nitroacetophenone, 1-5-bromo-2-hydroxy-3-nitrophenyl ethan-1-one, ethanone,1-5-bromo-2-hydroxy-3-nitrophenyl, ethanone, 1-5-bromo-2-hydroxy-3-nitrophenyl, 3'-bromo-6'-hydroxy-5'-nitroacetophenone, 1-acetyl-5-bromo-2-hydroxy-3-nitrobenzene
PubChem CID 689071
IUPAC Name 1-(5-bromo-2-hydroxy-3-nitrophényl)éthane-1-one
SMILES CC(=O)C1=CC(Br)=CC(=C1O)[N+]([O-])=O

Spécifications

Melting Point 131°C
Quantity 5 g
Formula Weight 260.04
Percent Purity ≥98.0% (GC,T)
Physical Form Poudre cristalline
Chemical Name or Material 5′-Bromo-2′-hydroxy-3′-nitroacetophenone