missing translation for 'onlineSavingsMsg'
Learn More

5'-Bromo-2'-hydroxy-3'-nitroacetophenone, 97%

Catalog No. AAH2733106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2733106 5 g
AAH2733114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2733106 Supplier Thermo Scientific Chemicals Supplier No. H2733106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 70978-54-0 | C8H6BrNO4 | 260.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70978-54-0
Molecular Formula C8H6BrNO4
Molecular Weight (g/mol) 260.04
MDL Number MFCD01631134
InChI Key CLNIBJASCGZXHH-UHFFFAOYSA-N
PubChem CID 689071
IUPAC Name 1-(5-bromo-2-hydroxy-3-nitrophenyl)ethan-1-one
SMILES CC(=O)C1=CC(Br)=CC(=C1O)[N+]([O-])=O

Specifications

Melting Point 129°C to 132°C
Quantity 5 g
Synonym 1-5-bromo-2-hydroxy-3-nitrophenyl ethanone, 5'-bromo-2'-hydroxy-3'-nitroacetophenone, 3-bromo-6-hydroxy-5-nitroacetophenone, 5'-bromo-3'-nitro-2'-hydroxyacetophenone, 5-bromo-2-hydroxy-3-nitroacetophenone, 1-5-bromo-2-hydroxy-3-nitrophenyl ethan-1-one, ethanone,1-5-bromo-2-hydroxy-3-nitrophenyl, ethanone, 1-5-bromo-2-hydroxy-3-nitrophenyl, 3'-bromo-6'-hydroxy-5'-nitroacetophenone, 1-acetyl-5-bromo-2-hydroxy-3-nitrobenzene
Formula Weight 260.05
Percent Purity 97%
Chemical Name or Material 5'-Bromo-2'-hydroxy-3'-nitroacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only