missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxy-3-methylpyridine, 98%, Thermo Scientific Chemicals

Catalog No. AAH6425322
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6425322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6425322 Supplier Thermo Scientific Chemicals Supplier No. H6425322
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89488-30-2
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD03427657
InChI Key HXPMRPRBABWPKL-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxy-3-methylpyridine, 5-bromo-3-methylpyridin-2-ol, 5-bromo-2-hydroxy-3-picoline, 5-bromo-3-methylpyridin-2 1h-one, 5-bromo-3-methyl-pyridin-2-ol, 5-bromo-3-methyl-2-pyridinol, 2-hydroxy-5-bromo-3-picoline, 2-hydroxy-3-methyl-5-bromopyridine, 5-bromo-3-methyl-2-pyridone, 2 1h-pyridinone, 5-bromo-3-methyl
PubChem CID 285436
IUPAC Name 5-bromo-3-methyl-1H-pyridin-2-one
SMILES CC1=CC(=CNC1=O)Br

Specifications

Melting Point 170°C to 174°C (literature)
Color White
Quantity 100 g
Beilstein 1447443
Formula Weight 188.02
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-hydroxy-3-methylpyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only