missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-furaldehyde 98.0+%, TCI America™
$102.03 - $298.41
Chemical Identifiers
| CAS | 1899-24-7 |
|---|---|
| Molecular Formula | C5H3BrO2 |
| Molecular Weight (g/mol) | 174.98 |
| MDL Number | MFCD00159501 |
| InChI Key | WJTFHWXMITZNHS-UHFFFAOYSA-N |
| Synonym | 5-bromo-2-furaldehyde, 5-bromofurfural, 5-bromo-2-furfural, 5-bromofuran-2-carboxaldehyde, 5-bromofurancarboxaldehyde, 2-furancarboxaldehyde, 5-bromo, 5-bromofuraldehyde, 5-bromo-furan-2-carbaldehyde, 2-furaldehyde, 5-bromo, 5-bromo-furfural |
| PubChem CID | 600328 |
| IUPAC Name | 5-bromofuran-2-carbaldehyde |
| SMILES | BrC1=CC=C(O1)C=O |
Chemical Identifiers
| 1899-24-7 | |
| 174.98 | |
| WJTFHWXMITZNHS-UHFFFAOYSA-N | |
| 600328 | |
| BrC1=CC=C(O1)C=O |
| C5H3BrO2 | |
| MFCD00159501 | |
| 5-bromo-2-furaldehyde, 5-bromofurfural, 5-bromo-2-furfural, 5-bromofuran-2-carboxaldehyde, 5-bromofurancarboxaldehyde, 2-furancarboxaldehyde, 5-bromo, 5-bromofuraldehyde, 5-bromo-furan-2-carbaldehyde, 2-furaldehyde, 5-bromo, 5-bromo-furfural | |
| 5-bromofuran-2-carbaldehyde |
Specifications
| 1899-24-7 | |
| 112°C | |
| MFCD00159501 | |
| 5-bromo-2-furaldehyde, 5-bromofurfural, 5-bromo-2-furfural, 5-bromofuran-2-carboxaldehyde, 5-bromofurancarboxaldehyde, 2-furancarboxaldehyde, 5-bromo, 5-bromofuraldehyde, 5-bromo-furan-2-carbaldehyde, 2-furaldehyde, 5-bromo, 5-bromo-furfural | |
| BrC1=CC=C(O1)C=O | |
| 174.98 | |
| 174.98 | |
| Crystalline Powder |
| 84°C | |
| C5H3BrO2 | |
| 5 g | |
| WJTFHWXMITZNHS-UHFFFAOYSA-N | |
| 5-bromofuran-2-carbaldehyde | |
| 600328 | |
| ≥98.0% (GC) | |
| 5-Bromo-2-furaldehyde |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator