missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluoropyrimidine 98.0+%, TCI America™

Catalog No. B36851G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B36851G Supplier TCI America Supplier No. B36851G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62802-38-4
Molecular Formula C4H2BrFN2
Molecular Weight (g/mol) 176.98
MDL Number MFCD07787364
InChI Key CTWZYPZCDJKBRS-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromopyrimidine, 5-bromo-2-fluoro-pyrimidine, pyrimidine, 5-bromo-2-fluoro, pubchem3589, acmc-1b67r, ksc352k7r, #, 5-bromanyl-2-fluoranyl-pyrimidine, abbypharma ap-11-1804
PubChem CID 600690
IUPAC Name 5-bromo-2-fluoropyrimidine
SMILES FC1=NC=C(Br)C=N1

Specifications

Melting Point 92°C
Color Yellow
Quantity 1 g
Formula Weight 176.98
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-fluoropyrimidine