missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluoropyridine, 98%

Catalog No. aal1953714
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAL1953714 25 g
AAL1953706 5 g
2 options

Catalog No. AAL1953714

Supplier: Thermo Scientific Chemicals L1953714

Only null left

CAS: 766-11-0 | C5H3BrFN | 175.988 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 766-11-0
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.988
MDL Number MFCD01863742
InChI Key MYUQKYGWKHTRPG-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromopyridine, 3-bromo-6-fluoropyridine, 5-bromo-2-fluoro pyridine, 5-bromo-2-fluoro-pyridine, pyridine, 5-bromo-2-fluoro, 2-fluoro-5-bromo pyridine, pubchem3550, 2-fluro-5-bromo-pyridine, acmc-209p4s, bromo-2-fluoropyridine, 5
PubChem CID 2783168
IUPAC Name 5-bromo-2-fluoropyridine
SMILES C1=CC(=NC=C1Br)F

Specifications

Density 1.72
Boiling Point 162°C to 164°C
Flash Point 73°C (163°F)
Odor Characteristic
Refractive Index 1.5294
Quantity 25 g
UN Number UN2810
Beilstein 1363171
Formula Weight 175.99
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-fluoropyridine
Show More Show Less
Hazard Category H227-H302+H332-H311-H315-H319-H335
Hazard Statement GHS H Statement
H311-H227-H302-H332-H315-H319-H335
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only