missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorobenzoic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 146328-85-0
Molecular Formula C7H4BrFO2
Molecular Weight (g/mol) 219
MDL Number MFCD00143423
InChI Key PEXAZYDITWXYNJ-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromobenzoic acid, 5-bromo-2-fluoro-benzoic acid, buttpark 19\01-67, benzoic acid, 5-bromo-2-fluoro, 3-bromo-6-fluoro-benzoic acid, pubchem1312, acmc-209cww, 3-bromo-6-fluorobenzoic acid, ksc183q6j, rarechem al bo 0757
PubChem CID 2736315
IUPAC Name 5-bromo-2-fluorobenzoic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC398590050
SDS View Documents
Thermo Scientific Chemicals
398590050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

146328-85-0
219
PEXAZYDITWXYNJ-UHFFFAOYSA-N
2736315
C1=CC(=C(C=C1Br)C(=O)O)F
C7H4BrFO2
MFCD00143423
2-fluoro-5-bromobenzoic acid, 5-bromo-2-fluoro-benzoic acid, buttpark 19\01-67, benzoic acid, 5-bromo-2-fluoro, 3-bromo-6-fluoro-benzoic acid, pubchem1312, acmc-209cww, 3-bromo-6-fluorobenzoic acid, ksc183q6j, rarechem al bo 0757
5-bromo-2-fluorobenzoic acid

Specifications

146328-85-0
100.0
Glass bottle
BrC6H3(F)COOH
5g
PEXAZYDITWXYNJ-UHFFFAOYSA-N
5-bromo-2-fluorobenzoic acid
2736315
97%
96.0
96% min. (HPLC)
C7H4BrFO2
MFCD00143423
2-fluoro-5-bromobenzoic acid, 5-bromo-2-fluoro-benzoic acid, buttpark 19\01-67, benzoic acid, 5-bromo-2-fluoro, 3-bromo-6-fluoro-benzoic acid, pubchem1312, acmc-209cww, 3-bromo-6-fluorobenzoic acid, ksc183q6j, rarechem al bo 0757
C1=CC(=C(C=C1Br)C(=O)O)F
219
219
5-Bromo-2-fluorobenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only