missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluoro-4-methylpyridine 98.0+%, TCI America™

Catalog No. B35791G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35791G 1 g
B35795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35791G Supplier TCI America Supplier No. B35791G
Only null left

Chemical Identifiers

CAS 864830-16-0
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD09839213
InChI Key PFODFMZCBQUYNG-UHFFFAOYSA-N
Synonym 5-bromo-2-fluoro-4-picoline, 5-bromo-2-fluoro-4-methyl-pyridine, 2-fluoro-4-methyl-5-bromopyridine, pubchem3547, acmc-209q9w, ksc495s8j
PubChem CID 23157062
IUPAC Name 5-bromo-2-fluoro-4-methylpyridine
SMILES CC1=CC(=NC=C1Br)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 190.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-fluoro-4-methylpyridine