missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-ethynylpyridine 98.0+%, TCI America™

Catalog No. p-7124001
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5179200MG 200 mg
B51791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5179200MG Supplier TCI America Supplier No. B5179200MG

Chemical Identifiers

CAS 111770-86-6
Molecular Formula C7H4BrN
Molecular Weight (g/mol) 182.02
MDL Number MFCD03095276
InChI Key VGLAJERHBBEPQP-UHFFFAOYSA-N
PubChem CID 13979734
IUPAC Name 5-bromo-2-ethynylpyridine
SMILES BrC1=CN=C(C=C1)C#C

Specifications

Melting Point 85°C
Quantity 200 mg
Formula Weight 182.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-ethynylpyridine