missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2'-deoxyuridine 98.0+%, TCI America™

Catalog No. p-7119130
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15751G 1 g
B15755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15751G Supplier TCI America Supplier No. B15751G
Only null left

Chemical Identifiers

CAS 59-14-3
Molecular Formula C9H11BrN2O5
Molecular Weight (g/mol) 307.10
MDL Number MFCD00006529
InChI Key WOVKYSAHUYNSMH-RRKCRQDMSA-N
Synonym 5-bromo-2'-deoxyuridine, broxuridine, bromodeoxyuridine, brdu, 5-bromodeoxyuridine, 5-brdu, budr, 5-bromouracil deoxyriboside, bromouracil deoxyriboside, 5-bromodesoxyuridine
PubChem CID 6035
ChEBI CHEBI:472552
IUPAC Name 5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O

Specifications

Color White
Quantity 1 g
Formula Weight 307.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2′-deoxyuridine
RTECSNumber YU7350000
TSCA Yes