missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2'-Deoxyuridine, >99%, MP Biomedicals™
$208.96
Chemical Identifiers
| CAS | 59-14-3 |
|---|---|
| Molecular Formula | C9H11BrN2O5 |
| Molecular Weight (g/mol) | 307.10 |
| MDL Number | MFCD00006529 |
| InChI Key | WOVKYSAHUYNSMH-RRKCRQDMSA-N |
| Synonym | 5-bromo-2'-deoxyuridine, broxuridine, bromodeoxyuridine, brdu, 5-bromodeoxyuridine, 5-brdu, budr, 5-bromouracil deoxyriboside, bromouracil deoxyriboside, 5-bromodesoxyuridine |
| PubChem CID | 6035 |
| ChEBI | CHEBI:472552 |
| IUPAC Name | 5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
| SMILES | OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN10017101
|
N/A | 1 g |
Each for $208.96
|
|
|||||
Chemical Identifiers
| 59-14-3 | |
| 307.10 | |
| WOVKYSAHUYNSMH-RRKCRQDMSA-N | |
| 6035 | |
| 5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
| C9H11BrN2O5 | |
| MFCD00006529 | |
| 5-bromo-2'-deoxyuridine, broxuridine, bromodeoxyuridine, brdu, 5-bromodeoxyuridine, 5-brdu, budr, 5-bromouracil deoxyriboside, bromouracil deoxyriboside, 5-bromodesoxyuridine | |
| CHEBI:472552 | |
| OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O |
Specifications
| 59-14-3 | |
| >99% | |
| MFCD00006529 | |
| WOVKYSAHUYNSMH-RRKCRQDMSA-N | |
| 5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione | |
| 6035 | |
| 307.1 | |
| 5-Bromo-2'-Deoxyuridine |
| White | |
| C9H11BrN2O5 | |
| 5-bromo-2'-deoxyuridine, broxuridine, bromodeoxyuridine, brdu, 5-bromodeoxyuridine, 5-brdu, budr, 5-bromouracil deoxyriboside, bromouracil deoxyriboside, 5-bromodesoxyuridine | |
| OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O | |
| 307.10 | |
| CHEBI:472552 | |
| Solid |
Safety and Handling
EINECSNumber : 200-415-9
Recommended Storage : Store at -20°C.