missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2'-deoxycytidine, MP Biomedicals™

5-Bromo-2'-deoxycytidine can be incorporated into DNA in the presence of DNA polymerase, replacing dCTP

Supplier:  MP Biomedicals Inc 0210016683

 View more versions of this product

Catalog No. ICN10016683

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5-Bromo-2'-deoxycytidine monohydrate
White
250 mg
KISUPFXQEHWGAR-RRKCRQDMSA-N
4-amino-5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
472175
Powder
1022-79-3
C9H12BrN3O4
5-bromo-2'-deoxycytidine, 5-bromodeoxycytidine, 4-amino-5-bromo-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, unii-6m3tp1aibh, brcdr, bromodeoxycytidine, 6m3tp1aibh, bcdr, 5-bromo-2'-deoxycytidine monohydrate, cytidine, 5-bromo-2'-deoxy
C1C(C(OC1N2C=C(C(=NC2=O)N)Br)CO)O
306.116
306.1 (Anhydrous)

Chemical Identifiers

1022-79-3
306.116
5-bromo-2'-deoxycytidine, 5-bromodeoxycytidine, 4-amino-5-bromo-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, unii-6m3tp1aibh, brcdr, bromodeoxycytidine, 6m3tp1aibh, bcdr, 5-bromo-2'-deoxycytidine monohydrate, cytidine, 5-bromo-2'-deoxy
4-amino-5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
C9H12BrN3O4
KISUPFXQEHWGAR-RRKCRQDMSA-N
472175
C1C(C(OC1N2C=C(C(=NC2=O)N)Br)CO)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 213-824-2

Recommended Storage : Store at 0°C. Store Desiccated.

SDS
Documents

Documents

Promotions

Promotions