missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2'-deoxycytidine, 99%

Catalog No. AAJ6545603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6545603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6545603 Supplier Thermo Scientific Chemicals Supplier No. J6545603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1022-79-3 | C9H12BrN3O4 | 306.116 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1022-79-3
Molecular Formula C9H12BrN3O4
Molecular Weight (g/mol) 306.116
MDL Number MFCD00047496
InChI Key KISUPFXQEHWGAR-RRKCRQDMSA-N
Synonym 5-bromo-2'-deoxycytidine, 5-bromodeoxycytidine, 4-amino-5-bromo-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, unii-6m3tp1aibh, brcdr, bromodeoxycytidine, 6m3tp1aibh, bcdr, 5-bromo-2'-deoxycytidine monohydrate, cytidine, 5-bromo-2'-deoxy
PubChem CID 472175
IUPAC Name 4-amino-5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES C1C(C(OC1N2C=C(C(=NC2=O)N)Br)CO)O

Specifications

Color White to Yellow
Quantity 1 g
Formula Weight 307.1
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 5-Bromo-2'-deoxycytidine
EINECSNumber 213-824-2
RTECSNumber HA3705000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only