missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-cyanopyridine 98.0+%, TCI America™

Catalog No. B36105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36105G 5 g
B361025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36105G Supplier TCI America Supplier No. B36105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 97483-77-7
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.008
MDL Number MFCD00234144
InChI Key DMSHUVBQFSNBBL-UHFFFAOYSA-N
Synonym 5-bromo-2-cyanopyridine, 5-bromopicolinonitrile, 5-bromo-2-pyridinecarbonitrile, 2-cyano-5-bromopyridine, 2-pyridinecarbonitrile, 5-bromo, 3-bromo-6-pyridinecarbonitrile, 5-bromo-pyridine-2-carbonitrile, 2-cyano-5-bromopyridin, pubchem2108, 5-bromo 2-cyanopyridine
PubChem CID 817154
IUPAC Name 5-bromopyridine-2-carbonitrile
SMILES C1=CC(=NC=C1Br)C#N

Specifications

Melting Point 131°C
Boiling Point 110°C
Quantity 5 g
UN Number 3439
Formula Weight 183.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-cyanopyridine