missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-cyanopyridine, 95%, Thermo Scientific Chemicals

$91.79 - $97.97

Chemical Identifiers

CAS 97483-77-7
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.008
MDL Number MFCD00234144
InChI Key DMSHUVBQFSNBBL-UHFFFAOYSA-N
Synonym 5-bromo-2-cyanopyridine, 5-bromopicolinonitrile, 5-bromo-2-pyridinecarbonitrile, 2-cyano-5-bromopyridine, 2-pyridinecarbonitrile, 5-bromo, 3-bromo-6-pyridinecarbonitrile, 5-bromo-pyridine-2-carbonitrile, 2-cyano-5-bromopyridin, pubchem2108, 5-bromo 2-cyanopyridine
PubChem CID 817154
IUPAC Name 5-bromopyridine-2-carbonitrile
SMILES C1=CC(=NC=C1Br)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1962103
View Documents
Thermo Scientific Chemicals
L1962103
1 g
Each for $91.79
Only null left
 
AAL1962106
View Documents
Thermo Scientific Chemicals
L1962106
5 g
Each for $97.97
Only null left
 
Description

Description

It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol

Notes
Store in cool place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

97483-77-7
183.008
DMSHUVBQFSNBBL-UHFFFAOYSA-N
817154
C1=CC(=NC=C1Br)C#N
C6H3BrN2
MFCD00234144
5-bromo-2-cyanopyridine, 5-bromopicolinonitrile, 5-bromo-2-pyridinecarbonitrile, 2-cyano-5-bromopyridine, 2-pyridinecarbonitrile, 5-bromo, 3-bromo-6-pyridinecarbonitrile, 5-bromo-pyridine-2-carbonitrile, 2-cyano-5-bromopyridin, pubchem2108, 5-bromo 2-cyanopyridine
5-bromopyridine-2-carbonitrile

Specifications

97483-77-7
C6H3BrN2
1 g
5-bromo-2-cyanopyridine, 5-bromopicolinonitrile, 5-bromo-2-pyridinecarbonitrile, 2-cyano-5-bromopyridine, 2-pyridinecarbonitrile, 5-bromo, 3-bromo-6-pyridinecarbonitrile, 5-bromo-pyridine-2-carbonitrile, 2-cyano-5-bromopyridin, pubchem2108, 5-bromo 2-cyanopyridine
DMSHUVBQFSNBBL-UHFFFAOYSA-N
5-bromopyridine-2-carbonitrile
817154
95%
124°C to 127°C
MFCD00234144
5498624
Soluble in dichloromethane,ether,ethyl acetate and methanol
C1=CC(=NC=C1Br)C#N
183.008
183.01
5-Bromo-2-cyanopyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only