missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-cyanopyridine, 95%, Thermo Scientific Chemicals

Catalog No. AAL1962103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1962103 1 g
AAL1962106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1962103 Supplier Thermo Scientific Chemicals Supplier No. L1962103

It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol

Notes
Store in cool place. Keep container tightly closed. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 97483-77-7
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.008
MDL Number MFCD00234144
InChI Key DMSHUVBQFSNBBL-UHFFFAOYSA-N
Synonym 5-bromo-2-cyanopyridine, 5-bromopicolinonitrile, 5-bromo-2-pyridinecarbonitrile, 2-cyano-5-bromopyridine, 2-pyridinecarbonitrile, 5-bromo, 3-bromo-6-pyridinecarbonitrile, 5-bromo-pyridine-2-carbonitrile, 2-cyano-5-bromopyridin, pubchem2108, 5-bromo 2-cyanopyridine
PubChem CID 817154
IUPAC Name 5-bromopyridine-2-carbonitrile
SMILES C1=CC(=NC=C1Br)C#N

Specifications

Melting Point 124°C to 127°C
Quantity 1 g
Beilstein 5498624
Solubility Information Soluble in dichloromethane,ether,ethyl acetate and methanol
Formula Weight 183.01
Percent Purity 95%
Chemical Name or Material 5-Bromo-2-cyanopyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only