missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloropyrimidine 97.0+%, TCI America™

Catalog No. p-7119800
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28471G 1 g
B28475G 5 g
B284725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B28471G Supplier TCI America Supplier No. B28471G
Only null left

Chemical Identifiers

CAS 32779-36-5
Molecular Formula C4H2BrClN2
Molecular Weight (g/mol) 193.43
MDL Number MFCD00483232
InChI Key XPGIBDJXEVAVTO-UHFFFAOYSA-N
Synonym 2-chloro-5-bromopyrimidine, 5-bromo-2-chloro-pyrimidine, 5-bromo-2-chloropyrimidne, pyrimidine, 5-bromo-2-chloro, 5-bromo-2-chloro-1,3-diazine, pubchem5159, zlchem 1210, acmc-209hvm, 5-bromo chloropyrimidine, 5-bromo-2-chloroprimidine
PubChem CID 606665
IUPAC Name 5-bromo-2-chloropyrimidine
SMILES ClC1=NC=C(Br)C=N1

Specifications

Melting Point 78°C
Color White-Yellow
Boiling Point 95°C
Quantity 1 g
Formula Weight 193.43
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-chloropyrimidine