missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-adamantanone 98.0+%, TCI America™
$195.73 - $691.57
Chemical Identifiers
| CAS | 20098-20-8 |
|---|---|
| Molecular Formula | C10H13BrO |
| Molecular Weight (g/mol) | 229.117 |
| MDL Number | MFCD01321097 |
| InChI Key | TXEWIOREYPSNRR-UHFFFAOYSA-N |
| PubChem CID | 590905 |
| IUPAC Name | 5-bromoadamantan-2-one |
| SMILES | C1C2CC3CC(C2)(CC1C3=O)Br |
Chemical Identifiers
| 20098-20-8 | |
| 229.117 | |
| TXEWIOREYPSNRR-UHFFFAOYSA-N | |
| 5-bromoadamantan-2-one |
| C10H13BrO | |
| MFCD01321097 | |
| 590905 | |
| C1C2CC3CC(C2)(CC1C3=O)Br |
Specifications
| 20098-20-8 | |
| White-Yellow | |
| MFCD01321097 | |
| TXEWIOREYPSNRR-UHFFFAOYSA-N | |
| 5-bromoadamantan-2-one | |
| 590905 | |
| ≥98.0% (GC) | |
| 5-Bromo-2-adamantanone |
| 153°C | |
| C10H13BrO | |
| 1 g | |
| C1C2CC3CC(C2)(CC1C3=O)Br | |
| 229.117 | |
| 229.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No