missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(4-morpholinyl)benzonitrile, 98%

Catalog No. AAH61376MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61376MD 250 mg
AAH6137603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH61376MD Supplier Thermo Scientific Chemicals Supplier No. H61376MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1105665-08-4 | C11H11BrN2O | 267.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1105665-08-4
Molecular Formula C11H11BrN2O
Molecular Weight (g/mol) 267.126
InChI Key IENSJQPUDLLAOI-UHFFFAOYSA-N
Synonym 5-bromo-2-morpholino benzonitrile, 5-bromo-2-morpholinobenzonitrile, 5-bromo-2-morpholin-4-yl benzonitrile
PubChem CID 59558545
IUPAC Name 5-bromo-2-morpholin-4-ylbenzonitrile
SMILES C1COCCN1C2=C(C=C(C=C2)Br)C#N

Specifications

Quantity 250 mg
UN Number UN3439
Formula Weight 267.13
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-(4-morpholinyl)benzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only