missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-dichloropyrimidine 97.0+%, TCI America™

Catalog No. p-7119785
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28235G 5 g
B282325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28235G Supplier TCI America Supplier No. B28235G

Chemical Identifiers

CAS 36082-50-5
Molecular Formula C4HBrCl2N2
Molecular Weight (g/mol) 227.87
InChI Key SIKXIUWKPGWBBF-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-bromopyrimidine, 2,4-dichloro-5-bromo pyrimidine, 5-bromo-2,4-dichloro-pyrimidine, pyrimidine, 5-bromo-2,4-dichloro, 5-bromo-2,6-dichloropyrimidine, 5-bromo-2,4-dichloro-1,3-diazine, 5-brom-2,4-dichlorpyrimidin, zlchem 380, pubchem3993, acmc-1ad68
PubChem CID 289973
IUPAC Name 5-bromo-2,4-dichloropyrimidine
SMILES C1=C(C(=NC(=N1)Cl)Cl)Br

Specifications

Color White-Yellow
Boiling Point 128°C
Quantity 5 g
UN Number 2923
Formula Weight 227.87
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Bromo-2,4-dichloropyrimidine