missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2,3-pyridinediol 98.0+%, TCI America™
$84.70 - $1026.11
Chemical Identifiers
| CAS | 34206-49-0 |
|---|---|
| Molecular Formula | C5H4BrNO2 |
| Molecular Weight (g/mol) | 190.00 |
| MDL Number | MFCD08276314 |
| InChI Key | KXBXUHWCODHRHK-UHFFFAOYSA-N |
| Synonym | 5-Bromo-2,3-dihydroxypyridine |
| PubChem CID | 12563083 |
| IUPAC Name | 5-bromo-3-hydroxy-1,2-dihydropyridin-2-one |
| SMILES | OC1=CC(Br)=CNC1=O |
Chemical Identifiers
| 34206-49-0 | |
| 190.00 | |
| KXBXUHWCODHRHK-UHFFFAOYSA-N | |
| 12563083 | |
| OC1=CC(Br)=CNC1=O |
| C5H4BrNO2 | |
| MFCD08276314 | |
| 5-Bromo-2,3-dihydroxypyridine | |
| 5-bromo-3-hydroxy-1,2-dihydropyridin-2-one |
Specifications
| 34206-49-0 | |
| White-Yellow | |
| MFCD08276314 | |
| 5-Bromo-2,3-dihydroxypyridine | |
| OC1=CC(Br)=CNC1=O | |
| 190.00 | |
| 190.00 | |
| Crystalline Powder |
| 249°C | |
| C5H4BrNO2 | |
| 1 g | |
| KXBXUHWCODHRHK-UHFFFAOYSA-N | |
| 5-bromo-3-hydroxy-1,2-dihydropyridin-2-one | |
| 12563083 | |
| ≥98.0% (GC,T) | |
| 5-Bromo-2,3-pyridinediol |
Safety and Handling
TSCA : No