missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,3-dihydrobenzofuran-7-carboxylic Acid 97.0+%, TCI America™

Catalog No. B17055G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17055G 5 g
B17051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17055G Supplier TCI America Supplier No. B17055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41177-72-4
Molecular Formula C9H7BrO3
Molecular Weight (g/mol) 243.06
MDL Number MFCD00191391
InChI Key LEBMKAXASFPSFA-UHFFFAOYSA-N
Synonym 5-bromo-2,3-dihydrobenzofuran-7-carboxylic acid, 5-bromo-2,3-dihydrobenzo b furan-7-carboxylic acid, 5-bromocoumaran-7-carboxylic acid, 5-bromo-2,3-dihydro-benzofuran-7-carboxylic acid, 7-benzofurancarboxylic acid, 5-bromo-2,3-dihydro, zlchem 1161, acmc-1aqu8, 5-bromo-2,3-dihydrobenzofuran-7-carboxylicacid, 5-bromanyl-2,3-dihydro-1-benzofuran-7-carboxylic acid
PubChem CID 2794797
IUPAC Name 5-bromo-2,3-dihydro-1-benzofuran-7-carboxylic acid
SMILES OC(=O)C1=CC(Br)=CC2=C1OCC2

Specifications

Melting Point 232°C
Quantity 5 g
Formula Weight 243.06
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2,3-dihydrobenzofuran-7-carboxylic Acid