missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,3'-bipyridine, 95%, Thermo Scientific™

Catalog No. AAH2788003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2788003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2788003 Supplier Thermo Scientific Chemicals Supplier No. H2788003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 774-53-8
Molecular Formula C10H7BrN2
Molecular Weight (g/mol) 235.08
MDL Number MFCD09037420
InChI Key ILJRIAPZWHYPMK-UHFFFAOYSA-N
Synonym 5-bromo-2,3'-bipyridine, 5-bromo-2,3' bipyridinyl, 5-bromo-2-pyridin-3-yl pyridine, 5-bromo-2,3'bipyridine, 5-bromo-2,3-bipyridine, 2,3'-bipyridine, 5-bromo, 5-bromo-2,3' bipyridine
PubChem CID 11390677
SMILES BrC1=CN=C(C=C1)C1=CN=CC=C1

Specifications

Melting Point 75°C to 78°C
Quantity 1 g
IUPAC Name 5-bromo-2,3'-bipyridine
Formula Weight 235.08
Percent Purity 95%
Chemical Name or Material 5-Bromo-2,3'-bipyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures