missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,2'-bithiophene-5'-carboxaldehyde 98.0+%, TCI America™

Catalog No. p-7119323
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18741G 1 g
B18745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18741G Supplier TCI America Supplier No. B18741G
Only null left

Chemical Identifiers

CAS 110046-60-1
Molecular Formula C9H5BrOS2
Molecular Weight (g/mol) 273.16
MDL Number MFCD01321134
InChI Key NTHMTYNJFSUBMF-UHFFFAOYSA-N
Synonym 5-Bromo-5′C-formyl-2,2′C-bithiophene
PubChem CID 11065610
IUPAC Name 5'-bromo-[2,2'-bithiophene]-5-carbaldehyde
SMILES BrC1=CC=C(S1)C1=CC=C(S1)C=O

Specifications

Melting Point 147°C
Quantity 1 g
Formula Weight 273.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2,2′-bithiophene-5′-carboxaldehyde