missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,1,3-benzothiadiazole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC09210DE

Quantity 5g

 Afficher plus de versions de ce produit

Numéro de catalogue. CC09210DE

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

Spécifications

Identifiants chimiques

1753-75-9
215.068
LLCRUZDFDGTAAN-UHFFFAOYSA-N
2776295
C1=CC2=NSN=C2C=C1Br
C6H3BrN2S
MFCD00460091
5-bromobenzo c 1,2,5 thiadiazole, 5-bromobenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole,5-bromo, 5-bromobenzo 1,2,5 thiadiazole, 2,1,3-benzothiadiazole, 5-bromo, pubchem12684, acmc-209ea1, amth068, 5-bromobenz-2,1,3-thiadiazole, 5-bromo-benzo 1,2,5 thiadiazole
5-bromo-2,1,3-benzothiadiazole

Spécifications

5-Bromo-2, 1, 3-benzothiadiazole
96.5
97%
5g
5-bromobenzo c 1,2,5 thiadiazole, 5-bromobenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole,5-bromo, 5-bromobenzo 1,2,5 thiadiazole, 2,1,3-benzothiadiazole, 5-bromo, pubchem12684, acmc-209ea1, amth068, 5-bromobenz-2,1,3-thiadiazole, 5-bromo-benzo 1,2,5 thiadiazole
C1=CC2=NSN=C2C=C1Br
215.068
215.07
1753-75-9
100.0
C6H3BrN2S
MFCD00460091
LLCRUZDFDGTAAN-UHFFFAOYSA-N
5-bromo-2,1,3-benzothiadiazole
2776295
97%
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

FDS
Documents

Documents

Promotions

Promotions