missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,1,3-benzothiadiazole, 97%, Thermo Scientific™

Catalog No. CC09210DA
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC09210DA 1g
CC09210DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC09210DA Supplier Thermo Scientific Chemicals Supplier No. CC09210DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1753-75-9
Molecular Formula C6H3BrN2S
Molecular Weight (g/mol) 215.068
MDL Number MFCD00460091
InChI Key LLCRUZDFDGTAAN-UHFFFAOYSA-N
Synonym 5-bromobenzo c 1,2,5 thiadiazole, 5-bromobenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole,5-bromo, 5-bromobenzo 1,2,5 thiadiazole, 2,1,3-benzothiadiazole, 5-bromo, pubchem12684, acmc-209ea1, amth068, 5-bromobenz-2,1,3-thiadiazole, 5-bromo-benzo 1,2,5 thiadiazole
PubChem CID 2776295
IUPAC Name 5-bromo-2,1,3-benzothiadiazole
SMILES C1=CC2=NSN=C2C=C1Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 215.07
Percent Purity 97%
Chemical Name or Material 5-Bromo-2, 1, 3-benzothiadiazole
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.