missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,1,3-benzothiadiazole, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1753-75-9
Molecular Formula C6H3BrN2S
Molecular Weight (g/mol) 215.068
MDL Number MFCD00460091
InChI Key LLCRUZDFDGTAAN-UHFFFAOYSA-N
Synonym 5-bromobenzo c 1,2,5 thiadiazole, 5-bromobenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole,5-bromo, 5-bromobenzo 1,2,5 thiadiazole, 2,1,3-benzothiadiazole, 5-bromo, pubchem12684, acmc-209ea1, amth068, 5-bromobenz-2,1,3-thiadiazole, 5-bromo-benzo 1,2,5 thiadiazole
PubChem CID 2776295
IUPAC Name 5-bromo-2,1,3-benzothiadiazole
SMILES C1=CC2=NSN=C2C=C1Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC09210DA
SDS View Documents
Thermo Scientific Chemicals
CC09210DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC09210DE
SDS View Documents
Thermo Scientific Chemicals
CC09210DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1753-75-9
215.068
LLCRUZDFDGTAAN-UHFFFAOYSA-N
2776295
C1=CC2=NSN=C2C=C1Br
C6H3BrN2S
MFCD00460091
5-bromobenzo c 1,2,5 thiadiazole, 5-bromobenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole,5-bromo, 5-bromobenzo 1,2,5 thiadiazole, 2,1,3-benzothiadiazole, 5-bromo, pubchem12684, acmc-209ea1, amth068, 5-bromobenz-2,1,3-thiadiazole, 5-bromo-benzo 1,2,5 thiadiazole
5-bromo-2,1,3-benzothiadiazole

Specifications

1753-75-9
100.0
Amber glass bottle
MFCD00460091
5-bromobenzo c 1,2,5 thiadiazole, 5-bromobenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole,5-bromo, 5-bromobenzo 1,2,5 thiadiazole, 2,1,3-benzothiadiazole, 5-bromo, pubchem12684, acmc-209ea1, amth068, 5-bromobenz-2,1,3-thiadiazole, 5-bromo-benzo 1,2,5 thiadiazole
C1=CC2=NSN=C2C=C1Br
215.068
215.07
5-Bromo-2, 1, 3-benzothiadiazole
96.5
97%
C6H3BrN2S
1g
LLCRUZDFDGTAAN-UHFFFAOYSA-N
5-bromo-2,1,3-benzothiadiazole
2776295
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'