missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1H-pyrrolo[2,3-b]pyridine 98.0+%, TCI America™

Catalog No. B35881G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35881G 1 g
B35885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35881G Supplier TCI America Supplier No. B35881G

Chemical Identifiers

CAS 183208-35-7
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.04
MDL Number MFCD06659677
InChI Key LPTVWZSQAIDCEB-UHFFFAOYSA-N
Synonym 5-bromo-7-azaindole, 5-bromo-1h-pyrrolo 2,3-b pyridine, 1h-pyrrolo 2,3-b pyridine, 5-bromo, 5-bromo azaindole, 3-bromo-7h-pyrrolo 2,3-b pyridine, 5-bromo-1h-pyrrolo 2 , 3-b pyridine, 5-bromopyrrolo 2,3-b pyridine, 5-bromo,7-azaindole
PubChem CID 10307932
IUPAC Name 5-bromo-1H-pyrrolo[2,3-b]pyridine
SMILES BrC1=CN=C2NC=CC2=C1

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 1 g
Formula Weight 197.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-1H-pyrrolo[2,3-b]pyridine