missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1H-indazole, 97%

Catalog No. AC456740050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456740050 5 g
AC456740250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456740050 Supplier Thermo Scientific Chemicals Supplier No. 456740050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 53857-57-1 | C7H5BrN2 | 197.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53857-57-1
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.04
MDL Number MFCD00839493,MFCD26227374
InChI Key STVHMYNPQCLUNJ-UHFFFAOYSA-N
Synonym 5-bromoindazole, 5-bromo-2h-indazole, 1h-indazole, 5-bromo, 5-bromo-1hindazole, chembl16425, 5-brom-1h-indazol, 5-bromoazaindole, 5-bromo indazole, 5-bromo-indazole
PubChem CID 761929
IUPAC Name 5-bromo-1H-indazole
SMILES BrC1=CC=C2NN=CC2=C1

Specifications

Melting Point 123.0°C to 124.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 5-Bromo-1H-indazole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiou

RUO – Research Use Only