missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1H-benzimidazole, 97%, Thermo Scientific™

Chemical Identifiers

CAS 4887-88-1
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.04
MDL Number MFCD00160001
InChI Key GEDVWGDBMPJNEV-UHFFFAOYSA-N
Synonym 5-bromo-1h-benzimidazole, 5-bromo-1h-benzo d imidazole, 5-bromo-1h-1,3-benzodiazole, 5-bromobenzimidazole, 1h-benzimidazole, 5-bromo, 5-bromo-3h-1,3-benzodiazole, 6-bromobenzimidazole, 5-bromo-benzimidazole, 6-bromo benzimidazole, pubchem9618
PubChem CID 785299
IUPAC Name 6-bromo-1H-benzimidazole
SMILES BrC1=CC=C2N=CNC2=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC45910DE
View Documents
N/A 5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC45910EA
View Documents
N/A 10g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

4887-88-1
197.04
GEDVWGDBMPJNEV-UHFFFAOYSA-N
785299
BrC1=CC=C2N=CNC2=C1
C7H5BrN2
MFCD00160001
5-bromo-1h-benzimidazole, 5-bromo-1h-benzo d imidazole, 5-bromo-1h-1,3-benzodiazole, 5-bromobenzimidazole, 1h-benzimidazole, 5-bromo, 5-bromo-3h-1,3-benzodiazole, 6-bromobenzimidazole, 5-bromo-benzimidazole, 6-bromo benzimidazole, pubchem9618
6-bromo-1H-benzimidazole

Specifications

4887-88-1
C7H5BrN2
5-bromo-1h-benzimidazole, 5-bromo-1h-benzo d imidazole, 5-bromo-1h-1,3-benzodiazole, 5-bromobenzimidazole, 1h-benzimidazole, 5-bromo, 5-bromo-3h-1,3-benzodiazole, 6-bromobenzimidazole, 5-bromo-benzimidazole, 6-bromo benzimidazole, pubchem9618
BrC1=CC=C2N=CNC2=C1
197.04
197.03
5-Bromo-1H-benzimidazole
97%
MFCD00160001
GEDVWGDBMPJNEV-UHFFFAOYSA-N
6-bromo-1H-benzimidazole
785299
97%
Videos
Safety and Handling

Safety and Handling

GHS P Statement Harmful if swallowed.

GHS P Statement IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Avoid contact during pregnancy/while nursing.

Warning

SDS
Documents

Documents