missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1H-benzimidazole, 97%

Catalog No. AC451670050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451670050 5 g
AC451670250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC451670050 Supplier Thermo Scientific Chemicals Supplier No. 451670050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4887-88-1 | C7H5BrN2 | 197.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4887-88-1
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.04
MDL Number MFCD00160001
InChI Key GEDVWGDBMPJNEV-UHFFFAOYSA-N
Synonym 5-bromo-1h-benzimidazole, 5-bromo-1h-benzo d imidazole, 5-bromo-1h-1,3-benzodiazole, 5-bromobenzimidazole, 1h-benzimidazole, 5-bromo, 5-bromo-3h-1,3-benzodiazole, 6-bromobenzimidazole, 5-bromo-benzimidazole, 6-bromo benzimidazole, pubchem9618
PubChem CID 785299
SMILES BrC1=CC=C2N=CNC2=C1

Specifications

Melting Point 130°C to 134°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
IUPAC Name 6-bromo-1H-benzimidazole
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 5-Bromo-1H-benzimidazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid contact during pregnancy/while nursing.

RUO – Research Use Only