missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-1-methylisatin 98.0+%, TCI America™
Supplier: TCI America B47795G
Specifications
| 5-Bromo-1-methylisatin | |
| 171°C | |
| C9H6BrNO2 | |
| 5 g | |
| GEEDYJPPYNIZLX-UHFFFAOYSA-N | |
| 5-bromo-1-methyl-2,3-dihydro-1H-indole-2,3-dione | |
| 16357 | |
| ≥98.0% (GC) |
| 2058-72-2 | |
| Red-Yellow | |
| MFCD00456313 | |
| 5-Bromo-1-methylindole-2,3-dione, 5-Bromo-1-methyl-2,3-indolinedione | |
| CN1C(=O)C(=O)C2=CC(Br)=CC=C12 | |
| 240.06 | |
| 240.06 | |
| Crystalline Powder |
Chemical Identifiers
| 2058-72-2 | |
| 240.06 | |
| GEEDYJPPYNIZLX-UHFFFAOYSA-N | |
| 16357 | |
| CN1C(=O)C(=O)C2=CC(Br)=CC=C12 |
| C9H6BrNO2 | |
| MFCD00456313 | |
| 5-Bromo-1-methylindole-2,3-dione, 5-Bromo-1-methyl-2,3-indolinedione | |
| 5-bromo-1-methyl-2,3-dihydro-1H-indole-2,3-dione |
Safety and Handling
RTECSNumber : NL7878000
TSCA : No