missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-methylimidazole, 97%

Catalog No. p-4813193
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC381040050 5 g
AC381040250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC381040050 Supplier Thermo Scientific Chemicals Supplier No. 381040050

CAS: 1003-21-0 | C4H5BrN2 | 161.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1003-21-0
Molecular Formula C4H5BrN2
Molecular Weight (g/mol) 161.00
MDL Number MFCD01632218
InChI Key HATLLUIOEIXWGD-UHFFFAOYSA-N
Synonym 5-bromo-1-methyl-1h-imidazole, 5-bromo-n-methylimidazole, pubchem8951, 1h-imidazole, 5-bromo-1-methyl, 5-bromo-1-methyl-imidazole, imidazole,5-bromo-1-methyl, 5-bromo-n-methyl-imidazole, 5-bromo-1-methyl imidazole, acmc-1c6k3, 1-methyl-5-bromo-1h-imidazole
PubChem CID 1515258
IUPAC Name 5-bromo-1-methylimidazole
SMILES CN1C=NC=C1Br

Specifications

Melting Point 40°C to 41°C
Boiling Point 120°C (15.0 mmHg)
Flash Point 82°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 161
Percent Purity 97%
Chemical Name or Material 5-Bromo-1-methylimidazole
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only