missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-1-indanone 98.0+%, TCI America™
$44.28 - $371.56
Chemical Identifiers
| CAS | 34598-49-7 |
|---|---|
| Molecular Formula | C9H7BrO |
| Molecular Weight (g/mol) | 211.058 |
| MDL Number | MFCD00082718 |
| InChI Key | KSONICAHAPRCMV-UHFFFAOYSA-N |
| Synonym | 5-bromo-1-indanone, 5-bromoindanone, 5-bromoindan-1-one, 5-bromo-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 5-bromo-2,3-dihydro, 1-indanone, 5-bromo, 5-bromo-indanone, 5-bromo indanone, zlchem 204, 5-bromo-1-oxoindane |
| PubChem CID | 520695 |
| IUPAC Name | 5-bromo-2,3-dihydroinden-1-one |
| SMILES | C1CC(=O)C2=C1C=C(C=C2)Br |
Chemical Identifiers
| 34598-49-7 | |
| 211.058 | |
| KSONICAHAPRCMV-UHFFFAOYSA-N | |
| 520695 | |
| C1CC(=O)C2=C1C=C(C=C2)Br |
| C9H7BrO | |
| MFCD00082718 | |
| 5-bromo-1-indanone, 5-bromoindanone, 5-bromoindan-1-one, 5-bromo-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 5-bromo-2,3-dihydro, 1-indanone, 5-bromo, 5-bromo-indanone, 5-bromo indanone, zlchem 204, 5-bromo-1-oxoindane | |
| 5-bromo-2,3-dihydroinden-1-one |
Specifications
| 34598-49-7 | |
| C9H7BrO | |
| 1 g | |
| KSONICAHAPRCMV-UHFFFAOYSA-N | |
| 5-bromo-2,3-dihydroinden-1-one | |
| 520695 | |
| ≥98.0% (GC) | |
| 5-Bromo-1-indanone |
| 127°C | |
| MFCD00082718 | |
| 5-bromo-1-indanone, 5-bromoindanone, 5-bromoindan-1-one, 5-bromo-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 5-bromo-2,3-dihydro, 1-indanone, 5-bromo, 5-bromo-indanone, 5-bromo indanone, zlchem 204, 5-bromo-1-oxoindane | |
| C1CC(=O)C2=C1C=C(C=C2)Br | |
| 211.058 | |
| 211.06 | |
| Crystalline Powder |
Safety and Handling
TSCA : No