missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-indanone 98.0+%, TCI America™

Catalog No. B23421G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23421G 1 g
B23425G 5 g
B234225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B23421G Supplier TCI America Supplier No. B23421G
Only null left

Chemical Identifiers

CAS 34598-49-7
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.058
MDL Number MFCD00082718
InChI Key KSONICAHAPRCMV-UHFFFAOYSA-N
Synonym 5-bromo-1-indanone, 5-bromoindanone, 5-bromoindan-1-one, 5-bromo-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 5-bromo-2,3-dihydro, 1-indanone, 5-bromo, 5-bromo-indanone, 5-bromo indanone, zlchem 204, 5-bromo-1-oxoindane
PubChem CID 520695
IUPAC Name 5-bromo-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C=C(C=C2)Br

Specifications

Melting Point 127°C
Quantity 1 g
Formula Weight 211.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-1-indanone