missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-indanone, 97%

Catalog No. AC309570050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC309570050 5 g Glass bottle
AC309570010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC309570050 Supplier Thermo Scientific Chemicals Supplier No. 309570050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34598-49-7 | C9H7BrO | 211.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34598-49-7
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.06
MDL Number MFCD00082718
InChI Key KSONICAHAPRCMV-UHFFFAOYSA-N
Synonym 5-bromo-1-indanone, 5-bromoindanone, 5-bromoindan-1-one, 5-bromo-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 5-bromo-2,3-dihydro, 1-indanone, 5-bromo, 5-bromo-indanone, 5-bromo indanone, zlchem 204, 5-bromo-1-oxoindane
PubChem CID 520695
IUPAC Name 5-bromo-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C=C(C=C2)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 126.0°C to 130.0°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 211.06
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 5-Bromo-1-indanone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

RUO – Research Use Only