missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-indanone, 97+%

Catalog No. AAB2570703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2570703 1 g
AAB2570706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2570703 Supplier Thermo Scientific Chemicals Supplier No. B2570703
Only null left

CAS: 34598-49-7 | C9H7BrO | 211.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34598-49-7
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.058
MDL Number MFCD00082718
InChI Key KSONICAHAPRCMV-UHFFFAOYSA-N
Synonym 5-bromo-1-indanone, 5-bromoindanone, 5-bromoindan-1-one, 5-bromo-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 5-bromo-2,3-dihydro, 1-indanone, 5-bromo, 5-bromo-indanone, 5-bromo indanone, zlchem 204, 5-bromo-1-oxoindane
PubChem CID 520695
IUPAC Name 5-bromo-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C=C(C=C2)Br

Specifications

Melting Point 125°C to 129°C
Quantity 1 g
Beilstein 2357199
Formula Weight 211.06
Percent Purity 98%
Chemical Name or Material 5-Bromo-1-indanone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only