missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1,4-benzodioxane, 95%

Catalog No. AAH27709MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27709MD 250 mg
AAH2770903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27709MD Supplier Thermo Scientific Chemicals Supplier No. H27709MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58328-39-5 | C8H7BrO2 | 215.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58328-39-5
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD03095029
InChI Key NGOZRIZXELGVHK-UHFFFAOYSA-N
Synonym 5-bromo-1,4-benzodioxane, 5-bromo-2,3-dihydro-1,4-benzodioxane, 5-bromo-2,3-dihydrobenzo b 1,4 dioxine, acmc-209m3q, 5-bromo-2,3-dihydrobenzo 1,4 dioxin, 5-bromo-2,3-dihydro 1,4 benzodioxine, 5-bromo-2,3-dihydro 1,4-benzodioxine, 1,4-benzodioxin, 5-bromo-2,3-dihydro
PubChem CID 18787268
IUPAC Name 5-bromo-2,3-dihydro-1,4-benzodioxine
SMILES C1COC2=C(O1)C=CC=C2Br

Specifications

Refractive Index 1.5955
Quantity 250 mg
Formula Weight 215.05
Percent Purity 95%
Chemical Name or Material 5-Bromo-1,4-benzodioxane
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only