missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1,3-dimethyl-2-pyridone, 97%

Catalog No. AAH5227903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5227903 1 g
AAH5227906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5227903 Supplier Thermo Scientific Chemicals Supplier No. H5227903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 51417-13-1 | C7H8BrNO | 202.051 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51417-13-1
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.051
MDL Number MFCD16658881
InChI Key UEHHXNZKBMALIY-UHFFFAOYSA-N
Synonym 5-bromo-1,3-dimethyl-2-pyridone, 5-bromo-1,3-dimethylpyridin-2 1h-one, 5-bromo-1,3-dimethyl-1h-pyridin-2-one, 2 1h-pyridinone, 5-bromo-1,3-dimethyl, 1,3-dimethyl-5-bromopyridine-2 1h-one, 5-bromo-1,3-dimethyl-1,2-dihydropyridin-2-one
PubChem CID 12272664
IUPAC Name 5-bromo-1,3-dimethylpyridin-2-one
SMILES CC1=CC(=CN(C1=O)C)Br

Specifications

Melting Point 105°C to 106°C
Quantity 1 g
Formula Weight 202.05
Percent Purity 97%
Chemical Name or Material 5-Bromo-1,3-dimethyl-2-pyridone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only