missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Boc-amino)-1-pentanol, 96%
CAS: 75178-90-4 | C10H21NO3 | 203.28 g/mol
$113.68 - $389.97
Chemical Identifiers
| CAS | 75178-90-4 |
|---|---|
| Molecular Formula | C10H21NO3 |
| Molecular Weight (g/mol) | 203.28 |
| MDL Number | MFCD01862955 |
| InChI Key | DDGNGFVNTZJMMZ-UHFFFAOYSA-N |
| Synonym | 5-boc-amino-1-pentanol, tert-butyl n-5-hydroxypentyl carbamate, tert-butyl 5-hydroxypentyl carbamate, 5-tert-butoxycarbonylamino-1-pentanol, n-5-hydroxypentyl carbamic acid tert-butyl ester, tert-butyl 5-hydroxypentylcarbamate, carbamic acid, 5-hydroxypentyl-, 1,1-dimethylethyl ester, n-boc-5-aminopentanol, 5-boc-amino 1-pentanol, acmc-209ox9 |
| PubChem CID | 4456136 |
| IUPAC Name | tert-butyl N-(5-hydroxypentyl)carbamate |
| SMILES | CC(C)(C)OC(=O)NCCCCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1764903
|
Thermo Scientific Chemicals
L1764903 |
1 g |
Each for $113.68
|
|
|||||
|
AAL1764906
|
Thermo Scientific Chemicals
L1764906 |
5 g |
Each for $389.97
|
|
|||||
Description
5-(Boc-amino)-1-pentanol may be used as non-limiting linkers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-(Boc-amino)-1-pentanol may be used as non-limiting linkers.
Solubility
Difficult to mix in water.
Notes
Moisture sensitive. Store in dry condition. Keep away from active metals, bases, oxidizing agents and heat.
Chemical Identifiers
| 75178-90-4 | |
| 203.28 | |
| DDGNGFVNTZJMMZ-UHFFFAOYSA-N | |
| 4456136 | |
| CC(C)(C)OC(=O)NCCCCCO |
| C10H21NO3 | |
| MFCD01862955 | |
| 5-boc-amino-1-pentanol, tert-butyl n-5-hydroxypentyl carbamate, tert-butyl 5-hydroxypentyl carbamate, 5-tert-butoxycarbonylamino-1-pentanol, n-5-hydroxypentyl carbamic acid tert-butyl ester, tert-butyl 5-hydroxypentylcarbamate, carbamic acid, 5-hydroxypentyl-, 1,1-dimethylethyl ester, n-boc-5-aminopentanol, 5-boc-amino 1-pentanol, acmc-209ox9 | |
| tert-butyl N-(5-hydroxypentyl)carbamate |
Specifications
| 75178-90-4 | |
| (CH3)3COCONH(CH2)5OH | |
| MFCD01862955 | |
| 5-boc-amino-1-pentanol, tert-butyl n-5-hydroxypentyl carbamate, tert-butyl 5-hydroxypentyl carbamate, 5-tert-butoxycarbonylamino-1-pentanol, n-5-hydroxypentyl carbamic acid tert-butyl ester, tert-butyl 5-hydroxypentylcarbamate, carbamic acid, 5-hydroxypentyl-, 1,1-dimethylethyl ester, n-boc-5-aminopentanol, 5-boc-amino 1-pentanol, acmc-209ox9 | |
| DDGNGFVNTZJMMZ-UHFFFAOYSA-N | |
| 1 | |
| 1 g | |
| >109°C (228°F) | |
| 203.28 | |
| 5-(Boc-amino)-1-pentanol |
| C10H21NO3 | |
| 1.456 | |
| 3603659 | |
| Difficult to mix in water. | |
| CC(C)(C)OC(=O)NCCCCCO | |
| tert-butyl N-(5-hydroxypentyl)carbamate | |
| 203.28 | |
| 4456136 | |
| 96% |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only