missing translation for 'onlineSavingsMsg'
Learn More

5-(Boc-amino)-1-pentanol, 96%

Catalog No. AAL1764903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1764903 1 g
AAL1764906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1764903 Supplier Thermo Scientific Chemicals Supplier No. L1764903

CAS: 75178-90-4 | C10H21NO3 | 203.28 g/mol

5-(Boc-amino)-1-pentanol may be used as non-limiting linkers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(Boc-amino)-1-pentanol may be used as non-limiting linkers.

Solubility
Difficult to mix in water.

Notes
Moisture sensitive. Store in dry condition. Keep away from active metals, bases, oxidizing agents and heat.

Chemical Identifiers

CAS 75178-90-4
Molecular Formula C10H21NO3
Molecular Weight (g/mol) 203.28
MDL Number MFCD01862955
InChI Key DDGNGFVNTZJMMZ-UHFFFAOYSA-N
Synonym 5-boc-amino-1-pentanol, tert-butyl n-5-hydroxypentyl carbamate, tert-butyl 5-hydroxypentyl carbamate, 5-tert-butoxycarbonylamino-1-pentanol, n-5-hydroxypentyl carbamic acid tert-butyl ester, tert-butyl 5-hydroxypentylcarbamate, carbamic acid, 5-hydroxypentyl-, 1,1-dimethylethyl ester, n-boc-5-aminopentanol, 5-boc-amino 1-pentanol, acmc-209ox9
PubChem CID 4456136
IUPAC Name tert-butyl N-(5-hydroxypentyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCCO

Specifications

Refractive Index 1.456
Linear Formula (CH3)3COCONH(CH2)5OH
Beilstein 3603659
Solubility Information Difficult to mix in water.
Density 1
Quantity 1 g
Flash Point >109°C (228°F)
Formula Weight 203.28
Percent Purity 96%
Chemical Name or Material 5-(Boc-amino)-1-pentanol
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only